missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid, Sigma-Aldrich™

Product Code. 15850791
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850791 500 mL 500mL
15840791 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15850791 Supplier Sigma Aldrich Supplier No. 8201700500
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-17-3

Specifications

Melting Point 12°C
Density 1.27 g/cm3 at 20°C
MDL Number MFCD00002585
Quantity 500 mL
Synonym Pyruvic acid, Pyruvic acid
Molecular Weight (g/mol) 88.06
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.