missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidinedithiocarbamate ammonium salt, >95%, MP Biomedicals™

Product Code. 11450821 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450821 100 g 100g
11440821 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450821 Supplier MP Biomedicals Supplier No. 0215647080
  • ONLINE EXCLUSIVE PRICE
    401.00 EUR
    381.15 EUR / 100g
    Save 19.85 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H8NS2-
Molecular Weight (g/mol) 146.246
MDL Number MFCD00012720
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
Synonym ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

Color White
Assay Percent Range >95%
Quantity 100 g
Formula Weight 164.29
Physical Form Crystalline Powder
Chemical Name or Material Pyrrolidinedithiocarbamate ammonium salt, Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.