Learn More
Pyrrolidine-1-dithiocarboxylic Acid Ammonium Salt, Sigma-Aldrich™
Product Code. 15821391
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821391 100 g 100g
15811391 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15821391 Supplier Sigma Aldrich Supplier No. 8211000100
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5108-96-3
Specifications
Melting Point 146°C to 151°C
MDL Number MFCD00012720
Quantity 100 g
Synonym Pyrrolidine-1-dithiocarboxylic acid ammonium salt, Ammonium-N,N-tetramethylenedithiocarbamate, N,N-Tetramethylenedithiocarbaminic acid ammoniumsalt, Pyrrolidine-1-dithiocarboxylic acid ammonium salt
Molecular Weight (g/mol) 164.29
Grade Research