Learn More
Lonza™ PYROGENT™-5000 Kinetic Turbidimetric LAL Assay Kits
Product Code. 11640331
Change view
Click to view available options
Quantity:
4500 Tests
Unit Size:
Pack of 4500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11640331 4500 Tests Pack of 4500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11640331 Supplier Lonza™ Supplier No. N688
 Restricted Item
Estimated Shipment: 18-01-2027
Detection of Gram-negative bacterial endotoxin. Lonza PYROGENT™-5000 Kinetic Turbidimetric LAL Assay Kits is ideal for water samples, large volume parenterals and water rinse from medical devices.
  • Ideal for large numbersof samples
  • Sample mixed with reconstituted LAL reagent in 96-well plate
  • Placed in incubating absorbance plate reader
  • Automatically monitored over time for appearance of turbidity
  • Presence of endotoxin lysate will begin to gel causing solution to become cloudy or turbid
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4500 Tests