Learn More
Lonza Pyrogen-free Test Tubes
Description
- Made from USP Type 1 flint borosilicate glass
- Contain less than 0.005 EU/ml endotoxin
- Recommended for use as reaction tubes in gel clot assays.
Specifications
Specifications
| Length (Metric) | 75 mm |
| Material | Borosilicate Glass |
| Closure Type | Screw Cap |
| Product Type | Test Tube |