Learn More
Pyrocatechol, Sigma-Aldrich™
Product Code. 15811741
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15811741 1 kg 1kg
15801741 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15811741 Supplier Sigma Aldrich Supplier No. 8222611000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120-80-9
Specifications
Melting Point 103°C to 105°C
Density 1.37 g/cm3 at 20°C
Molecular Formula C6H4-1,2-(OH)2
MDL Number MFCD00002188
Quantity 1 kg
Synonym Pyrocatechol, 1,2-Dihydroxybenzene, Catechol, Pyrocatechol
Molecular Weight (g/mol) 110.11
Grade Research