missing translation for 'onlineSavingsMsg'
Learn More

Pyrocatechol Monoethyl Ether, Sigma-Aldrich™

Product Code. 15808680
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15808680 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808680 Supplier Sigma Aldrich Supplier No. 8019170250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 94-71-3
Melting Point 24°C to 28°C
Density 1.10 g/cm3 at 20°C
Boiling Point 214°C to 216°C/1013 hPa
Molecular Formula 2-(C2H5O)C6H4OH
MDL Number MFCD00002187
Quantity 250 mL
Synonym Pyrocatechol monoethyl ether, Guaethol, 2-Ethoxyphenol, Pyrocatechol monoethyl ether
Molecular Weight (g/mol) 138.16
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.