Learn More
Pyridostigmine-d6 bromide, MedChemExpress
Product Code. 16478418
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478418 10 mg 10mg
16432089 1 mg 1mg
16400828 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16478418 Supplier Medchem Express Supplier No. HYB0207AS
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
MedChemExpress Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
Specifications
Chemical Name or Material Pyridostigmine-d6 bromide
CAS 2375858-08-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C9H7D6BrN2O2
Color White
Solubility Information DMSO : 50 mg/mL (187.16 mM; Need ultrasonic)
SMILES O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])OC1=CC=C[N+](C)=C1.[Br-]
Molecular Weight (g/mol) 267.15
Quantity 10 mg
Formula Weight 267.15
Percent Purity 98.0%
Grade Research