Learn More
Pyridine-4-carboxaldoxime, 97%
Product Code. 11433617
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433617 100 g 100g
11423617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433617 Supplier Thermo Scientific Alfa Aesar Supplier No. A17371.22
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 696-54-8 | C6H6N2O | 122.13 g/mol
It is employed as an intermediate for bis(hydroxyiminomethylpyridinium)butenes and as reactivators of chlorpyrifos-inhibited acetylcholinesterase. It is also used in the chromatographic separation and NMR characterization of the isomers of 1,1?-Methylenebis{4-[(hydroxyimino)methyl]pyridinium) dichloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an intermediate for bis(hydroxyiminomethylpyridinium)butenes and as reactivators of chlorpyrifos-inhibited acetylcholinesterase. It is also used in the chromatographic separation and NMR characterization of the isomers of 1,1′-Methylenebis{4-[(hydroxyimino)methyl]pyridinium) dichloride.
Solubility
Soluble in water at 20°C 10 g/L. Soluble in DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 696-54-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006446
InChI Key OFYLBLSSPQTTHT-VMPITWQZSA-N
Synonym pyridine-4-aldoxime, 4-pyridinealdoxime, isonicotinaldehyde oxime, 4-pyridinecarboxaldehyde, oxime, isonicotinaldoxime, 4-hydroxyiminomethyl pyridine, 4-pyridinaldoxime, 4-pyridine aldoxime, pyridine, 4-formyl-, oxime, ccris 7744
PubChem CID 5324569
IUPAC Name 4-(nitrosomethylidene)-1H-pyridine
SMILES O\N=C\C1=CC=NC=C1
Specifications
Melting Point 130°C to 133°C
Quantity 100 g
Beilstein 2079
Solubility Information Soluble in water at 20°C 10g/L. Soluble in DMSO and methanol.
Formula Weight 122.13
Percent Purity 97%
Chemical Name or Material Pyridine-4-carboxaldoxime