missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 11314805
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314805 250 mg 250mg
11304805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314805 Supplier Thermo Scientific Alfa Aesar Supplier No. L17854.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181219-01-2
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.06
MDL Number MFCD01319051
InChI Key NLTIETZTDSJANS-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester
PubChem CID 2734648
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=NC=C1

Specifications

Melting Point 150°C to 152°C
Quantity 250 mg
Beilstein 7919059
Solubility Information Insoluble in water.
Formula Weight 205.07
Percent Purity 98%
Chemical Name or Material Pyridine-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.