Learn More
Pyridaben-d13, TRC
Specifications
Specifications
| Chemical Name or Material | Pyridaben D13 (tert-butylphenyl D13) |
| CAS | 2468638-05-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 D13 H12 Cl N2 O S |
| Quantity | 1 mg |
| Synonym | Pyridaben D13 (tert-butylphenyl D13) |
| InChI Formula | InChI=1S/C19H25ClN2OS/c1-18(2,3)14-9-7-13(8-10-14)12-24-15-11-21-22(19(4,5)6)17(23)16(15)20/h7-11H,12H2,1-6H3/i1D3,2D3,3D3,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1CSC2=C(Cl)C(=O)N(N=C2)C(C)(C)C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2-tert-butyl-4-chloro-5-[[2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenyl]methylsulfanyl]pyridazin-3-one |
| Molecular Weight (g/mol) | 378.013 |
| Show More |