Learn More
Azlon™ Labware Detergent
Product Code. 12549965
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12549965 5 L Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12549965 Supplier Azlon™ Supplier No. 40023
  • ONLINE EXCLUSIVE PRICE
    114.00 EUR
    85.87 EUR / Pack of 2
    Save 28.13 EUR
    24% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Contains no chlorine bleaches, phosphates, or enzymes. With excellent cleaning performance, Azlon™ Labware Detergent is ideal for use with laboratory glassware, plasticware, rubber, ceramics and stainless steel. Economically priced, non-corrosive, non-irritant, and totally biodegradable surface-active cleaning agent.
  • Excellent cleaning performance, ideal for use with laboratory glassware, plasticware, rubber, ceramics and stainless steel
  • Economical price 
  • Non-corrosive, non-irritant, and totally biodegradable surface-active cleaning agent
  • Free of chlorine bleaches, phosphates and enzymes
Specifications
Product Type Lipsol Detergent
Form Liquid
pH 10.78 (at 20°C)
Appearance Clear
Boiling Point 100°C
For Use With (Application) Ideal for use with laboratory Glassware, Plasticware, Rubber, Ceramics and Stainless Steel
Quantity 2 Pk.
Volume (English) 169.07oz.
Volume (Metric) 5 L