Learn More
PYREX™ Borosilicate Glass Graduated Cylinders
Product Code. 12672042
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Pack of 2
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
12672042 500 mL 5 mL Pack of 2
12682042 1000 mL 10 mL Pack of 2
12652042 100 mL 1 mL Pack of 2
10043927 250 mL 2 mL Pack of 2
12642042 50 mL 1 mL Pack of 2
15527710 2000 mL 20 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12672042 Supplier Pyrex Supplier No. 3280500
  • ONLINE EXCLUSIVE PRICE
    179.00 EUR
    154.34 EUR / Pack of 2
    Save 24.66 EUR
    13% Off
Estimated Shipment: 17-03-2027
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
  • Hexagonal base delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Specifications
Tolerance ±5 mL
Material Borosilicate Glass
Certifications/Compliance BS 604, DIN 12680, ISO 4788
Class Class B
Capacity (Metric) 500 mL
Height (Metric) 365 mm
Scale Blue enamel
Graduations 5 mL
Base Type Hexagonal
Scale Color Blue
Product Type Graduated Cylinder