Learn More
Pyrex™ Borosilicate Glass Cylinders in Squat Format
Product Code. 12682032
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
12682032 25 mL 1 mL Pack of 2
12632042 1000 mL 20 mL Pack of 2
12602042 100 mL 2 mL Pack of 2
12612042 250 mL 5 mL Pack of 2
12622042 500 mL 10 mL Pack of 2
12692032 50 mL 1 mL Pack of 2
12672032 10 mL 1 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12682032 Supplier Pyrex Supplier No. 328525
Estimated Shipment: 21-01-2027
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Squat Form are ideal for use in fume hoods and restricted spaces. Borosilicate glass gives superior chemical resistance.
  • Hexagonal base and squat form delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Specifications
Tolerance ±0.5 mL
Material Borosilicate Glass
Certifications/Compliance BS 604, DIN 12680, ISO 4788
Class Class B
Capacity (Metric) 25 mL
For Use With (Application) Confined Spaces
Height (Metric) 118 mm
Scale Blue enamel
Graduations 1 mL
Base Type Hexagonal
Scale Color Blue
Shape Round
Product Type Graduated Cylinder