Learn More
Pyrex™ Borosilicate Glass Cylinders in Squat Format
Product Code. 12622042
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
12622042 500 mL 10 mL Pack of 2
12632042 1000 mL 20 mL Pack of 2
12602042 100 mL 2 mL Pack of 2
12612042 250 mL 5 mL Pack of 2
12682032 25 mL 1 mL Pack of 2
12692032 50 mL 1 mL Pack of 2
12672032 10 mL 1 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12622042 Supplier Pyrex Supplier No. 3285500
  • ONLINE EXCLUSIVE PRICE
    179.00 EUR
    154.34 EUR / Pack of 2
    Save 24.66 EUR
    13% Off
Estimated Shipment: 22-12-2026
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Squat Form are ideal for use in fume hoods and restricted spaces. Borosilicate glass gives superior chemical resistance.
  • Hexagonal base and squat form delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Specifications
Tolerance ±5 mL
Material Borosilicate Glass
Certifications/Compliance BS 604, DIN 12680, ISO 4788
Class Class B
Capacity (Metric) 500 mL
For Use With (Application) Confined Spaces
Height (Metric) 254 mm
Scale Blue enamel
Graduations 10 mL
Base Type Hexagonal
Scale Color Blue
Shape Round
Product Type Graduated Cylinder