Learn More
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Product Code. 11902619
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
125 mL
150 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
3000 mL
5 L
Height (Metric):
52 mm
60 mm
70 mm
85 mm
105 mm
117 mm
118 mm
131 mm
145 mm
156 mm
180 mm
220 mm
280 mm
310 mm
365 mm
Diameter (Metric) Outer:
105 mm
131 mm
166 mm
187 mm
220 mm
25 mm
30 mm
42 mm
51 mm
64 mm
74 mm
79 mm
85 mm
87 mm
Body: 69 mm
Unit Size:
Each
Pack of 10
Pack of 2
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Diameter (Metric) Outer Unit Size
11902619 250 mL 145 mm 85 mm Pack of 10
10348669 200 mL 131 mm 79 mm Pack of 10
10460160 3000 mL 310 mm 187 mm Pack of 2
10765332 5 L 365 mm 220 mm Each
11972568 25 mL 70 mm 42 mm Pack of 10
11962568 100 mL 105 mm 64 mm Pack of 10
11961566 2000 mL 280 mm 166 mm Pack of 10
11982568 50 mL 85 mm 51 mm Pack of 10
12579105 5 mL 52 mm 25 mm Pack of 10
12589105 10 mL 60 mm 30 mm Pack of 10
11912619 500 mL 180 mm 105 mm Pack of 10
11922619 1000 mL 220 mm 131 mm Pack of 10
11910905 150 mL 118 mm 74 mm Pack of 10
12519115 300 mL 156 mm 87 mm Pack of 10
15685767 125 mL 117 mm Body: 69 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 11902619 Supplier Pyrex Supplier No. 1130/14D
  • ONLINE EXCLUSIVE PRICE
    73.25 EUR
    63.55 EUR / Pack of 10
    Save 9.70 EUR
    13% Off
View all promotions
Estimated Shipment: 10-12-2026
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
  • Complies with ISO 1773 and DIN 12353
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
Specifications
Certifications/Compliance ISO 1773, DIN 12353
Capacity (Metric) 250 mL
Material Borosilicate Glass
Graduations Yes
Height (Metric) 145 mm
Shape Conical
Writing Surface Yes
Graduated Yes
Diameter (Metric) Outer 85 mm
Closure Size 29