Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Product Code. 12375263
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
29/32
Height (Metric):
107 mm
151 mm
160 mm
198 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Height (Metric) Unit Size
12375263 1000 mL 29/32 198 mm Each
11921025 100 mL 29/32 107 mm Each
12041672 250 mL 29/32 151 mm Each
12071672 500 mL 29/32 160 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12375263 Supplier Pyrex™ Supplier No. FD1L/4RE
  • ONLINE EXCLUSIVE PRICE
    70.40 EUR
    61.09 EUR / Pack of 1
    Save 9.31 EUR
    13% Off
Estimated Shipment: 30-12-2026
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 198 mm
Standard Taper Joint 29/32