Learn More
Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes
Product Code. 15137193
Change view
Click to view available options
Capacity (Metric):
15 mL
45 mL
90 mL
170 mL
320 mL
600 mL
Diameter (Metric):
50 mm
60 mm
80 mm
95 mm
115 mm
140 mm
Height (Metric):
25 mm
30 mm
45 mm
55 mm
65 mm
80 mm
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Height (Metric) unitSize
15137193 320 mL 115 mm 65 mm Each
12053323 15 mL 50 mm 25 mm Pack of 10
12524267 320 mL 115 mm 65 mm Pack of 10
11357844 170 mL 95 mm 55 mm Pack of 10
15127193 170 mL 95 mm 55 mm Each
10528000 45 mL 60 mm 30 mm Pack of 10
12063323 90 mL 80 mm 45 mm Pack of 10
12683042 600 mL 140 mm 80 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15137193 missing translation for 'mfr' Pyrex missing translation for 'supplierNo' 1450/10D
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Manufactured from Pyrex™ borosilicate glass and complies to DIN 12336 standards. Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes are highly resistant to chemical erosion and thermal shock. Flat base with curved sides and spout provide easy handling and helps to prevent spillage.
  • Complies to DIN 12336
  • Manufactured from Pyrex™ borosilicate glass
  • High resistance to chemical erosion and thermal shock
  • Flat base
  • Curved sides with spout
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Capacity (Metric) 320 mL
Material Borosilicate Glass
Shape Round
Type Evaporating Dish
Height (Metric) 65 mm
Diameter (Metric) 115 mm