Learn More
- Home
- Products
- Flasks
- Erlenmeyer Flasks
- Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Product Code. 10793032
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Height (Metric):
57 mm
68 mm
76 mm
85 mm
105 mm
130 mm
160 mm
210 mm
280 mm
315 mm
Diameter (Metric) Outer:
25 mm
30 mm
42 mm
51 mm
64 mm
85 mm
105 mm
131 mm
166 mm
187 mm
Closure Size:
15
22
30
42
Unit Size:
Each
Pack of 10
Pack of 20
Pack of 5
Pack of 50
13 product options available for selection
| Product Code. | Capacity (Metric) | Height (Metric) | Diameter (Metric) Outer | Closure Size | Unit Size |
|---|---|---|---|---|---|
| 10793032 | 1000 mL | 210 mm | 131 mm | 42 | Each |
| 12588866 | 2000 mL | 280 mm | 166 mm | 42 | Pack of 5 |
| 11950905 | 250 mL | 130 mm | 85 mm | 42 | Pack of 10 |
| 10029219 | 250 mL | 130 mm | 85 mm | 42 | Each |
| 11981518 | 25 mL | 76 mm | 42 mm | 22 | Pack of 10 |
| 11920905 | 50 mL | 85 mm | 51 mm | 22 | Pack of 10 |
| 11940905 | 100 mL | 105 mm | 64 mm | 30 | Pack of 10 |
| 10358829 | 100 mL | 105 mm | 64 mm | 30 | Each |
| 11960905 | 500 mL | 160 mm | 105 mm | 42 | Pack of 10 |
| 15537670 | 10 mL | 68 mm | 30 mm | 15 | Pack of 50 |
| 15547670 | 3000 mL | 315 mm | 187 mm | 42 | Pack of 20 |
| 11991518 | 3000 mL | 315 mm | 187 mm | 42 | Pack of 5 |
| 15527670 | 5 mL | 57 mm | 25 mm | 15 | Pack of 50 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
This item is not returnable.
View return policy
Product Code.
10793032
Supplier
Pyrex™
Supplier No.
1137/10M
-
58.65 EUR
50.85 EUR / Pack of 1
Save 7.80 EUR
13% Off
Estimated Shipment: 03-03-2027
Log in to see stock.
Log in to see stock.
Zum Warenkorb hinzufügen
50.84 EUR valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
- PBT screw caps with PTFE faced sealing disc
- Erlenmeyer pattern
- Manufactured from Pyrex™ Borosilicate Glass
- Highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
- Graduated to full capacity
Spezifikation
Spezifikation
| Closure Material | SVL™ Phenolic, PTFE faced with Rubber disc |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 210 mm |
| Shape | Conical |
| Graduated | Yes |
| Closure Type | Screw Cap |
| Diameter (Metric) Outer | 131 mm |
| Closure Size | 42 |
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?