Learn More
Pyrene, Sigma-Aldrich™
Product Code. 15826680
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15826680 5 g 5g
15811361 50 g 50g
15821361 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15826680 Supplier Sigma Aldrich Supplier No. 8210510005
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 129-00-0
Specifications
Melting Point 145.3°C
Density 1.209 g/cm3 at 20°C
Boiling Point 393°C/1013 hPa
MDL Number MFCD00004136
Quantity 5 g
Synonym Pyrene, Benzo[def]phenanthrene, Pyrene
Molecular Weight (g/mol) 202.25
Grade Research