Learn More
Pyoluteorin, TRC
Product Code. 18032140
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032140 25 mg 25mg
18082010 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18032140 Supplier Toronto Research Chemicals Supplier No. TRCP84037525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25683-07-2
Molecular Formula C11 H7 Cl2 N O3
Molecular Weight (g/mol) 272.0842
Synonym Ketone, 4,5-dichloropyrrol-2-yl 2,6-dihydroxyphenyl (7CI,8CI), Pyoluteorin (6CI), (4,5-Dichloro-1H-pyrrol-2-yl)(2,6-dihydroxyphenyl)methanone, 4,5-Dichloropyrrol-2-yl 2,6-dihydroxyphenylketone, NSC 143092
IUPAC Name (4,5-dichloro-1H-pyrrol-2-yl)-(2,6-dihydroxyphenyl)methanone
SMILES Oc1cccc(O)c1C(=O)c2cc(Cl)c(Cl)[nH]2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H7Cl2NO3/c12-5-4-6(14-11(5)13)10(17)9-7(15)2-1-3-8(9)16/h1-4,14-16H
Formula Weight 270.9803
Percent Purity >95%
Chemical Name or Material Pyoluteorin
Grade HPLC