missing translation for 'onlineSavingsMsg'
Learn More

PX-478, MedChemExpress

Product Code. 16496457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496457 5 mg 5mg
16453728 10 mg 10mg
16410117 50 mg 50mg
16484796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496457 Supplier Medchem Express Supplier No. HY10231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PX-478 is an orally active HIF-1α inhibitor with potent antitumor activities. PX-478 can cross the blood-brain barrier.

PX-478 is an orally active HIF-1α inhibitor with potent antitumor activities. PX-478 can cross the blood-brain barrier.

Specifications

Chemical Name or Material PX-478
Physical Form Solid
CAS 685898-44-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H20Cl4N2O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (253.73 mM; Need ultrasonic) ∣H2O : ≥ 35 mg/mL (88.81 mM)
SMILES [O-][N+](CCCl)(CCCl)C(C=C1)=CC=C1C[C@H](N)C(O)=O.Cl.Cl
Molecular Weight (g/mol) 394.12
Formula Weight 394.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.