Learn More
Clark Scientific™ PVC Large White Spill Trays
Product Code. 12988837
Change view
Click to view available options
Type:
Large Spill Tray
Small Spill Tray
Size:
475 x 360 mm
700 x 450 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Size Unit Size
12988837 Large Spill Tray 700 x 450 mm Each
12968837 Small Spill Tray 475 x 360 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12988837 Supplier Clark Scientific™ Supplier No. WPVC large
  • ONLINE EXCLUSIVE PRICE
    53.15 EUR
    48.25 EUR / Pack of 1
    Save 4.90 EUR
    9% Off
Estimated Shipment: 22-12-2026
Manufactured from rigid PVC, these large white plastic spill trays are for use outside radiation areas.
    • Manufactured from rigid PVC
    • Available in yellow for use in radiation areas
    • Available in white for use outside radiation areas
    • Colourless PVC liners are available
Specifications
Color White
Material PVC
Type Large Spill Tray
For Use With (Application) Outside radiation areas
Packaging Quantity 1 Ea.
Size 700 x 450 mm
Length (Metric) 700 mm
Height (Metric) 450 mm