Learn More
Fisherbrand™ PVC Tubing
Product Code. 12375869 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
16 mm
19 mm
Unit Size:
10m
Product Code. 12375869
Only null left
Add to basket
Add to basket
Suitable for moderate pressure or vacuum applications
All dimensions are approximate.
  • Transparent, colorless
  • Terylene mesh reinforced wall
  • Flexible yet great inherent strength
  • Suitable for moderate pressure or vacuum applications
Specifications
Diameter (Metric) Inner 16 mm
Material PVC
Overall Length (Metric) 10 m
For Use With (Application) Suitable For Moderate Pressure Or Vacuum Applications
Color Colorless