Learn More
Navailles PVC Stool with Foot Control
Product Code. 17456162
Change view
Click to view available options
Seating Height:
460 to 590 mm
570 to 670 mm
Includes:
Seat, Backrest, Foot Control
Seat, Foot Control
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Seating Height Includes unitSize
17456162 460 to 590 mm Seat, Backrest, Foot Control Each
17486162 570 to 670 mm Seat, Foot Control Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17456162 Supplier Navailles Supplier No. 4GM 37GPLU 90 737
  • ONLINE EXCLUSIVE PRICE
    466.20 EUR
    422.00 EUR / Pack of 1
    Save 44.20 EUR
    9% Off
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
The PVC stool with Foot Control offers the advantages of being compact, polyvalent, easily adjustable, and hygienic.
  • Chrome, 5-branch, reinforced, scuff-resistant base
  • Castors with rubber wheels
  • M1 vinyl round seat with high-density soft foam
  • Chrome gas-lift column with height adjustment and shock absorber
  • Foot control for adjusting height to keep hands clean
Specifications
Color Gray
Arm Style No Armrests
Certifications/Compliance M1
For Use With (Application) Medical Practices
Includes Seat, Backrest, Foot Control
Load Bearing Capacity (English) 265 lb.
Load Bearing Capacity (Metric) 120 kg
Seating Height 460 to 590 mm
Width (English) 23.2 in.
Width (Metric) 59 cm