Learn More
Brady™ PVC Cable
Product Code. 11711116 Shop All Brady Products
Change view
Click to view available options
Color:
Red/Silver
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11711116 Red/Silver Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11711116 Supplier Brady™ Supplier No. 805844
  • ONLINE EXCLUSIVE PRICE
    121.00 EUR
    115.70 EUR / Pack of 1
    Save 5.30 EUR
    4% Off
In Stock
Flexible steel wire with protective PVC coating for use with a padlock.
  • Flexible steel wire with protective PVC coating for use with a padlock.
Specifications
Product Type Padlock Cable
Color Red/Silver
Description Cobra Cable
Diameter (English) 0.31 in.
Diameter (Metric) 8 mm
Length (English) 6.56 ft.
Length (Metric) 2 m
Quantity 1 Ea.