missing translation for 'onlineSavingsMsg'
Learn More

Putrescine, ∽98%, MP Biomedicals™

Product Code. 11483120 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483120 1 g 1g
11493120 5 g 5g
11403130 25 g 25g
11413130 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11483120 Supplier MP Biomedicals Supplier No. 0210044101
  • ONLINE EXCLUSIVE PRICE
    66.15 EUR
    63.00 EUR / 1g
    Save 3.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Putrescine is a GABA precursor in many biological systems and synthon for amido-ureas

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-60-1
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008235
InChI Key KIDHWZJUCRJVML-UHFFFAOYSA-N
Synonym 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina
PubChem CID 1045
ChEBI CHEBI:17148
IUPAC Name butane-1,4-diamine
SMILES C(CCN)CN

Specifications

Color Colorless
Density 0.877g/mL at 25°C
Assay Percent Range ∽98%
Quantity 1 g
Formula Weight 88.15
Physical Form Liquid
Chemical Name or Material Putrescine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.