Learn More
PURUS™ Frames for Biohybrid™ Contamination Control Mats
Product Code. 15372002 Shop All Purus Products
Change view
Click to view available options
Dimensions (L x W):
965 x 965 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15372002 965 x 965 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15372002 Supplier PURUS™ Supplier No. PF 3838 01 W 00
  • ONLINE EXCLUSIVE PRICE
    240.41 EUR
    158.96 EUR / Pack of 1
    Save 81.45 EUR
    33% Off
Estimated Shipment: 04-02-2027
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS&trade Biohybrid&trade mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications
Dimensions (L x W) 965 x 965 mm
For Use With (Application) For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
For Use With (Equipment) 914 x 914 mm PURUS™ Biohybrid™ Mats
Thickness (Metric) 3.175 mm
Color White
Material Polystyrene