missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ PuroSorb™ PAD Macroporous Adsorbents

Product Code. 17049583
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Product Line:
PAD1200
PAD610
PAD950
Unit Size:
1kg
250g
5kg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Line unitSize
17049583 250 g PAD950 250g
17009583 1 kg PAD1200 1kg
17019583 250 g PAD610 250g
17029583 5 kg PAD610 5kg
17039583 1 kg PAD610 1kg
17059583 5 kg PAD950 5kg
17069583 1 kg PAD950 1kg
17089573 250 g PAD1200 250g
17099573 5 kg PAD1200 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17049583 Supplier Purolite™ Supplier No. LS07380192

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PuroSorb™ adsorbents are macroporous, rigid, highly cross-linked synthetic polymers. They have a variety of base matrices, porosities (or pore volumes), surface areas, and pore sizes. They also exhibit non-polar or hydrophobic tendencies so they adsorb hydrophobic organic compounds.

  • Stable in acidic or alkaline solutions and in most organic solvents
  • Regenerate under mild conditions for prolonged life
  • Chemically stable, insoluble
  • Spherical beads
  • Macroporous structure
  • Made of styrene, DVB or methacrylate
  • Stevia Purification
  • Peptide extraction
  • Decolorization
  • Macrolide purification
  • Compliant with ResAP(2004) 3
  • Halal and TSE/BSE/GMO free
  • Compliant with FDA regulation 21 CFR 173.65

Specifications

Final Product Type White to off-white spherical beads
Product Line PAD950
Product Type Macroporous Polymethacrylic Adsorbent
Quantity 250 g
pH Range 1 to 14 pH
Particle Size 350 to 1200 μm
Content And Storage Use within 5 years from date of manufacture, store at 2°C to 20°C
Format Wet
Moisture 65 to 76%
Porosity 120 Å
Form Wet
Matrix Adsorbent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.