Learn More
MedChemExpress Pulchinenoside C
Specifications
Specifications
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Molecular Weight (g/mol) | 1221.38 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 129741-57-7 |
| Product Type | Natural Products |
| Purity | 98.0% |
| Solubility | DMSO : 125 mg/mL (102.34 mM∣Need ultrasonic) |
| Synonym | Anemoside B4 |
| Quantity | 10 mg |