Learn More
Buerkle™ Puerckhauer Soil Sampler
Product Code. 11892303 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
81 cm
117.5 cm
Diameter (Metric) Outer Bottom:
20 mm
25 mm
Diameter (Metric) Outer Top:
20 mm
28 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Outer Bottom Diameter (Metric) Outer Top Unit Size
11892303 117.5 cm 25 mm 28 mm Each
10618898 81 cm 20 mm 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11892303 Supplier Buerkle™ Supplier No. 53505018
  • ONLINE EXCLUSIVE PRICE
    1105.00 EUR
    961.35 EUR / Pack of 1
    Save 143.65 EUR
    13% Off
Estimated Shipment: 23-10-2026
Puerckhauer soil sampler for heavy and stony soils.
Very robust Puerckhauer-sampler for heavy and stony soils. Model 5350-5018 has a conically-tapered 60 cm tip which makes it much easier to extract the driven-in sampler. Ideal for N-min sampling 0–90 cm.
  • Depth graduation 10 cm divisions
  • Attachable handle, with non-slip plastic grips
  • Reinforced impact head
  • Stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Puerckhauer Soil Sampler
Material Stainless Steel
Includes Attachable handle, with non-slip Plastic grips
Graduated Yes
For Use With (Application) For Heavy and Stony Soils, 0 to 90 cm N-Min Sampling
Weight (Metric) 3800 g
Diameter (Metric) Outer Bottom 25 mm
Diameter (Metric) Outer Top 28 mm
Length (Metric) 117.5 cm