Learn More
Chemsplash PU Grip Anti-Slip Sole OverShoe
Product Code. 30058269
Change view
Click to view available options
Size:
Universal
Unit Size:
200 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058269 Universal 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058269 Supplier Chemsplash Supplier No. 2782.400.007
  • ONLINE EXCLUSIVE PRICE
    345.06 EUR
    292.26 EUR / 200 pairs
    Save 52.80 EUR
    15% Off
Estimated Shipment: 04-01-2027
The Grip Overshoe with Anti-Slip sole is made from Chemsplash 55 gsm micro laminate fabric.
Chemsplash Grip Overshoe
  • 55GSM Type PB 6B Material
  • 70gsm Grey PU Anti-Slip Sole
  • Elasticated Top and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Grip Overshoe with Anti-Slip sole is made from Chemsplash 55gsm Micro Laminate fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This overshoe is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforming to ISO 14644-1:2015 Cleanroom Class 5-9, it features a 70gsm PU Anti-Slip Sole that is also anti-static. Chemsplash accessories are designed to protect the wearer from the environment or vice versa, providing maximum comfort and wearability.
Specifications
Size Universal
Material Microporous Composite Fabric, Proprietary Laminated Non-woven Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)