Learn More
Macherey-Nagel™ PTFE Thread Tape
Product Code. 10378341 Shop All Macherey-Nagel Products
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10378341 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10378341 Supplier Macherey-Nagel™ Supplier No. 706512
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
Estimated Shipment: 25-11-2026
Specifications
For Use With (Application) For Sealing
Length (Metric) 12 m
Material PTFE
Product Type Thread Tape
Width (Metric) 12 mm