Learn More
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Product Code. 11898862 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
8 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
75 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
10 mm
13 mm
Unit Size:
Each
Pack of 10
Pack of 5
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11898862 60 mm 10 mm Pack of 10
10018131 60 mm 10 mm Each
11888962 13 mm 8 mm Pack of 10
12641156 50 mm 8 mm Pack of 5
11537812 45 mm 8 mm Pack of 5
11888862 45 mm 8 mm Pack of 10
12379507 15 mm 4.5 mm Pack of 5
16265811 70 mm 10 mm Pack of 10
10373592 70 mm 10 mm Each
12661156 30 mm 6 mm Pack of 10
11527792 30 mm 6 mm Pack of 5
12359507 12 mm 6 mm Pack of 5
12651156 12 mm 6 mm Pack of 10
11517812 35 mm 6 mm Pack of 5
11868862 35 mm 6 mm Pack of 10
11868962 75 mm 13 mm Pack of 5
11527812 40 mm 8 mm Pack of 5
11878862 40 mm 8 mm Pack of 10
11808972 25 mm 10 mm Pack of 10
11507812 25 mm 6 mm Pack of 5
11848862 25 mm 6 mm Pack of 10
11838862 20 mm 6 mm Pack of 10
11597802 20 mm 6 mm Pack of 5
11898962 15 mm 8 mm Pack of 10
11587802 12 mm 4.5 mm Pack of 5
11818862 12 mm 4.5 mm Pack of 10
11808862 8 mm 3 mm Pack of 10
12349507 8 mm 3 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Product Code. 11898862
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
Provides stable stirring action. FisherbrandTM PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action.
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 10 mm
Length (Metric) 60 mm
Shape Cylinder, Pivot ring