Learn More
- Home
- Products
- Hotplates and Stirrers
- Stirring Bars and Rods
- Fisherbrand™ PTFE Stir Bars with Pivot Ring
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Product Code. 11848862 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
8 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
75 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
10 mm
13 mm
Unit Size:
Each
Pack of 10
Pack of 5
28 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) | Unit Size |
|---|---|---|---|
| 11848862 | 25 mm | 6 mm | Pack of 10 |
| 10018131 | 60 mm | 10 mm | Each |
| 11898862 | 60 mm | 10 mm | Pack of 10 |
| 11888962 | 13 mm | 8 mm | Pack of 10 |
| 12641156 | 50 mm | 8 mm | Pack of 5 |
| 11537812 | 45 mm | 8 mm | Pack of 5 |
| 11888862 | 45 mm | 8 mm | Pack of 10 |
| 12379507 | 15 mm | 4.5 mm | Pack of 5 |
| 16265811 | 70 mm | 10 mm | Pack of 10 |
| 10373592 | 70 mm | 10 mm | Each |
| 12661156 | 30 mm | 6 mm | Pack of 10 |
| 11527792 | 30 mm | 6 mm | Pack of 5 |
| 12359507 | 12 mm | 6 mm | Pack of 5 |
| 12651156 | 12 mm | 6 mm | Pack of 10 |
| 11517812 | 35 mm | 6 mm | Pack of 5 |
| 11868862 | 35 mm | 6 mm | Pack of 10 |
| 11868962 | 75 mm | 13 mm | Pack of 5 |
| 11527812 | 40 mm | 8 mm | Pack of 5 |
| 11878862 | 40 mm | 8 mm | Pack of 10 |
| 11808972 | 25 mm | 10 mm | Pack of 10 |
| 11507812 | 25 mm | 6 mm | Pack of 5 |
| 11838862 | 20 mm | 6 mm | Pack of 10 |
| 11597802 | 20 mm | 6 mm | Pack of 5 |
| 11898962 | 15 mm | 8 mm | Pack of 10 |
| 11587802 | 12 mm | 4.5 mm | Pack of 5 |
| 11818862 | 12 mm | 4.5 mm | Pack of 10 |
| 11808862 | 8 mm | 3 mm | Pack of 10 |
| 12349507 | 8 mm | 3 mm | Pack of 5 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
Product Code.
11848862
Spotlight Offers
Use promo code 28038
Use promo code 28038
Estimated Shipment: 29-12-2026
Log in to see stock.
Log in to see stock.
Add to basket
8.01 EUR valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 25 mm |
| Shape | Cylinder, Pivot ring |