Learn More
BRAND™ Straight-Bore PTFE Stopcocks
Product Code. 10360142 Shop All Brand Products
Change view
Click to view available options
Fits Tubing ID:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fits Tubing ID Unit Size
10360142 9 mm Each
10300102 8 mm Each
10304901 7 mm Each
10203541 6 mm Each
10152011 5 mm Each
10374991 3 mm Each
10569341 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10360142 Supplier BRAND™ Supplier No. 88640
Estimated Shipment: 21-12-2026
PTFE construction with polypropylene valve lever. Provides outstanding chemical resistance. Includes integrated hose nozzles.
  • PTFE construction
  • Polypropylene valve
  • With integrated one-way hose nozzle
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 5 mm
Material PTFE
Fits Tubing ID 9 mm
Includes Integrated hose nozzle
Plug Material Polypropylene