Learn More
Masterflex™ PTFE 3-Way Direct Lift Solenoid Valve
Product Code. 11700029
Change view
Click to view available options
Voltage:
12 VDC
24 VDC
Maximum Pressure:
18 psi
30 psi
60 psi
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Maximum Pressure unitSize
11700029 12 VDC 30 psi Each
11790019 24 VDC 60 psi Each
12843223 24 VDC 30 psi Each
13257499 24 VDC 18 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11700029 Supplier Masterflex™ Supplier No. 0154011
  • ONLINE EXCLUSIVE PRICE
    242.00 EUR
    193.97 EUR / Pack of 1
    Save 48.03 EUR
    19% Off
Estimated Shipment: 08-10-2026
Specifications
For Use With (Application) Ideal for use with high-purity or corrosive fluids
Max. Operating Temperature (Metric) 70°C
Material PTFE
Quantity 1 Each
Type 3-way Direct Lift Solenoid Valve
Voltage 12 VDC
Maximum Pressure 30 psi