Learn More
Hamilton PT1000 Probe Support Stand
Product Code. 31257792
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
31257792 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31257792 Supplier Hamilton Supplier No. 10176378
  • ONLINE EXCLUSIVE PRICE
    243.00 EUR
    220.66 EUR / Pack of 1
    Save 22.34 EUR
    9% Off
Estimated Shipment: 25-11-2026
Provides a stable and secure platform for the PT1000 temperature probe when used with a magnetic heater stirrer. Ensures reliable positioning during laboratory temperature monitoring.
  • Adjustable PT1000 temperature probe support stand
  • Attaches securely to the MagHeat magnetic stirrer
  • Keeps the temperature probe stable during operation
Specifications
Type Temperature Probe
Warranty None
Material Stainless Steel