Learn More
IKA™ PT 1000.70 3.3 Borosilicate Glass Temperature Sensor
Product Code. 10024053 Shop All IKA Products
Change view
Click to view available options
Diameter (Metric):
7 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10024053 7 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10024053 Supplier IKA™ Supplier No. 0003736000
  • ONLINE EXCLUSIVE PRICE
    215.00 EUR
    186.00 EUR / Pack of 1
    Save 29.00 EUR
    13% Off
In Stock
Specifications
Diameter (Metric) 7 mm
For Use With (Equipment) For 3589W/6097C
Model PT 1000.70
Temperature (Metric) -10°C to +400°C
Type Temperature Sensor