missing translation for 'onlineSavingsMsg'
Learn More

PSI-697, MedChemExpress

Product Code. 16482687
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482687 50 mg 50mg
16456598 10 mg 10mg
16449167 5 mg 5mg
16431529 1 mg 1mg
16416796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16482687 Supplier Medchem Express Supplier No. HY15526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PSI-697 is an oral P-selectin inhibitor with an IC50 of 125 μM.

PSI-697 is an oral P-selectin inhibitor with an IC50 of 125 μM.

Specifications

Chemical Name or Material PSI-697
CAS 851546-61-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18ClNO3
Quantity 50 mg
Synonym P-Selectin Inhibitor
Solubility Information DMSO : ≥ 45.8 mg/mL (124.51 mM)
SMILES O=C(C1=C(O)C(CC2=CC=C(Cl)C=C2)=NC3=C(CCCC4)C4=CC=C13)O
Molecular Weight (g/mol) 367.83
Formula Weight 367.83
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.