missing translation for 'onlineSavingsMsg'
Learn More

PSG11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

149.35 EUR - 422.30 EUR

Specifications

Antigen PSG11
Concentration 0.19 mg/mL
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16805954
View Documents
Proteintech
16352-1-AP-20UL
20 μL
149.35 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16895944
View Documents
Proteintech
16352-1-AP-150UL
150 μL
422.30 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregcy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.
Specifications

Specifications

PSG11
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse
Q9UQ72
5680
PSG11 Fusion Protein Ag9562
Primary
-20°C
PSG11
0.19 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
PS beta G 11, PS beta G 13, PSBG 11, PSBG 13, PSG11, PSG13, PSG14
PSG11
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.