Learn More
PSG11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16352-1-AP-150UL
This item is not returnable.
View return policy
Description
PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregcy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.Specifications
| PSG11 | |
| Polyclonal | |
| Unconjugated | |
| PSG11 | |
| PS beta G 11, PS beta G 13, PSBG 11, PSBG 13, PSG11, PSG13, PSG14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5680 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UQ72 | |
| PSG11 | |
| PSG11 Fusion Protein Ag9562 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |