Learn More
Pseudomethylionone, TRC
Product Code. 17952995
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952995 250 mg 250mg
17903745 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17952995 Supplier Toronto Research Chemicals Supplier No. TRCP839535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26651-96-7
Molecular Formula C14 H22 O
Molecular Weight (g/mol) 206.3239
Synonym 7,11-Dimethyl-4,6,10-dodecatrien-3-one, Methylpseudoionone, Pseudoionone, n-methyl-, Pseudomethylionone
IUPAC Name (4E,6E)-7,11-dimethyldodeca-4,6,10-trien-3-one
SMILES CCC(=O)\C=C\C=C(/C)\CCC=C(C)C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C14H22O/c1-5-14(15)11-7-10-13(4)9-6-8-12(2)3/h7-8,10-11H,5-6,9H2,1-4H3/b11-7+,13-10+
Formula Weight 206.1671
Percent Purity >95%
Chemical Name or Material Pseudomethylionone
Grade HPLC