CAS RN 26651-96-7
CAS RN 26651-96-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pseudomethylionone, TRC
CAS: 26651-96-7 Molecular Formula: C14 H22 O Molecular Weight (g/mol): 206.3239 Synonym: 7,11-Dimethyl-4,6,10-dodecatrien-3-one,Methylpseudoionone,Pseudoionone, n-methyl-,Pseudomethylionone IUPAC Name: (4E,6E)-7,11-dimethyldodeca-4,6,10-trien-3-one SMILES: CCC(=O)\C=C\C=C(/C)\CCC=C(C)C
2
7,11-Dimethyl-4,6,10-dodecatrien-3-one 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality