missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSEM 89S
GREENER_CHOICE

Product Code. 18798736
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18798736 5 mg 5mg
18769406 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18798736 Supplier Tocris Bioscience™ Supplier No. 6426/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 chimeric ion channel agonist

PSEM 89S is a PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 chimeric ion channel agonist (EC50 values are 3.4 and 2.2 μM, respectively). Activates cortical neurons expressing PSAML141F,Y115F-5-HT3 chimeric ion channels and inhibits activity of neurons expressing PSAML141F-GlyR in vitro. Silences hypothalamic AGRP neurons transfected with PSAML141F-GlyR chimeric ion channels in mice in vivo, leading to suppression of feeding. Brain penetrant.

Plasmid vectors for the transfection of cells with PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 are available from Addgene.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3 S)-1-Azabicyclo[2.2.2 ]oct-3-yl-2,5-dimethoxybenzamide trifluoroacetate
CAS 1336913-03-1
Quantity 5 mg
Target Pharmacologically Selective Effector Molecules/PSEM
Molecular Formula C16H22N2O3.CF3CO2H
Formula Weight 404.38
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.