missing translation for 'onlineSavingsMsg'
Learn More

Prucalopride Succinate, TRC

Product Code. 30010160
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010160 50 mg 50mg
30004375 500 mg 500mg
30006467 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010160 Supplier Toronto Research Chemicals Supplier No. TRCP83895050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179474-85-2
Molecular Formula C18 H26 Cl N3 O3 . C4 H6 O4
Molecular Weight (g/mol) 485.96
Synonym Butanedioic acid, compd. with 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-7-benzofurancarboxamide (1:1), 7-Benzofurancarboxamide, 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-, butanedioate (1:1) (9CI), Prucalopride succinate, R 108512, Resolor, Resotran
IUPAC Name 4-amino-5-chloro-N-[1-(3-methoxypropyl)piperidin-4-yl]-2,3-dihydro-1-benzofuran-7-carboxamide;butanedioic acid
SMILES COCCCN1CCC(CC1)NC(=O)c2cc(Cl)c(N)c3CCOc23.OC(=O)CCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H26ClN3O3.C4H6O4/c1-24-9-2-6-22-7-3-12(4-8-22)21-18(23)14-11-15(19)16(20)13-5-10-25-17(13)14;5-3(6)1-2-4(7)8/h11-12H,2-10,20H2,1H3,(H,21,23);1-2H2,(H,5,6)(H,7,8)
Formula Weight 485.1929
Percent Purity >95%
Chemical Name or Material Prucalopride Succinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.