Learn More
Prucalopride Succinate, TRC
Product Code. 30004375
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004375 500 mg 500mg
30006467 100 mg 100mg
30010160 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004375 Supplier Toronto Research Chemicals Supplier No. TRCP838950500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 179474-85-2
Molecular Formula C18 H26 Cl N3 O3 . C4 H6 O4
Molecular Weight (g/mol) 485.96
Synonym Butanedioic acid, compd. with 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-7-benzofurancarboxamide (1:1), 7-Benzofurancarboxamide, 4-amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-, butanedioate (1:1) (9CI), Prucalopride succinate, R 108512, Resolor, Resotran
IUPAC Name 4-amino-5-chloro-N-[1-(3-methoxypropyl)piperidin-4-yl]-2,3-dihydro-1-benzofuran-7-carboxamide;butanedioic acid
SMILES COCCCN1CCC(CC1)NC(=O)c2cc(Cl)c(N)c3CCOc23.OC(=O)CCC(=O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H26ClN3O3.C4H6O4/c1-24-9-2-6-22-7-3-12(4-8-22)21-18(23)14-11-15(19)16(20)13-5-10-25-17(13)14;5-3(6)1-2-4(7)8/h11-12H,2-10,20H2,1H3,(H,21,23);1-2H2,(H,5,6)(H,7,8)
Formula Weight 485.1929
Percent Purity >95%
Chemical Name or Material Prucalopride Succinate
Grade HPLC