missing translation for 'onlineSavingsMsg'
Learn More

Prucalopride succinate, MedChemExpress

Product Code. 16491497
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491497 50 mg 50mg
16461046 200 mg 200mg
16405956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491497 Supplier Medchem Express Supplier No. HY12694
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Prucalopride succinate is a selective, high affinity 5-HT4 receptor agonist with pKi of 8.6/8.1 for 5-HT4a/4b.

Prucalopride succinate is a selective, high affinity 5-HT4 receptor agonist with pKi of 8.6/8.1 for 5-HT4a/4b.

Specifications

Chemical Name or Material Prucalopride succinate
CAS 179474-85-2
Color White
Purity Grade Notes Research
Molecular Formula C22H32ClN3O7
Quantity 50 mg
Synonym R-108512
Solubility Information DMSO : 50 mg/mL (102.89 mM; Need ultrasonic)
SMILES O=C(O)CCC(O)=O.O=C(C1=C(OCC2)C2=C(N)C(Cl)=C1)NC3CCN(CCCOC)CC3
Molecular Weight (g/mol) 485.96
Formula Weight 485.96
Percent Purity 99.83%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.