Learn More
Hamilton™ PRP-1 Semiprep/Preparative Guard Column Starter Kit (1 holder, 1 cartridge), Stainless Steel
Product Code. 11573394
Product Code. 11573394 Supplier Hamilton™ Supplier No. 79121
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Guard columns can provide many benefits when used upstream from an analytical, preparative, or semi-preparative column. Guard columns are typically chosen to have the same packing as the primary column. In addition to protective uses, guard columns can be used as sacrificial preparative components.
Specification Sheet
  • Separation Mode – Reversed-Phase
  • Stationary Phase – PRP-1
  • Format – Guard Cartridge Kit
  • Hardware Inner Diameter – 4.6 mm
  • Hardware Length – 20 mm
  • Particle Size – 12-20 μm
  • Packing Material Type – PSDVB
  • Guard Column Size – Semiprep/Preparative
  • Hardware Material – Stainless Steel
  • Max Pressure – 5,000 psi
  • Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
  • Compound Classification – Organics
  • UOM – 1 holder, 1 cartridge
Specifications
Particle Size 12 to 20 μm
Length (Metric) 20 mm
Type PRP-1 Guard Columns
Diameter (Metric) Inner 4.6 mm