Learn More
Tocris Bioscience™ ProtiFect STAR™ Protein Transfection Reagent
Product Code. 30000661
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000661 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30000661 Supplier Tocris Bioscience™ Supplier No. 8098/1
Protein transfection reagent
ProtiFect STAR™ Protein Transfection Reagent is a transfection reagent capable of delivering a wide range of proteins with varying molecular weights (up to 240 kDa) and isoelectric points to the cytoplasm, including Cas9 ribonucleoproteins and BSA-FITC. ProtiFect STAR™ is a gemini amphiphile, which bypasses endocytic pathways. Can be used in vivo and in vitro in a variety of cell types including primary cells, T-cells, B-cells and stem cells. For more information on how to use ProtiFect STAR™, please see our protocol .
This compound is supplied pre-dissolved in ethanol (10 mg/mL).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Target Transfection Reagent